1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride

A2495-5G
TCA2495-5GEA 21.32 USD
TCA2495-5G
1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride ≥98.0% (by HPLC, total nitrogen)
1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride

CAS Number: 130198-05-9
MDL Number: MFCD06658143
Molecular Formula: C15H23NO2
Molecular Weight: 285.81
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 176

Formula: C₁₅H₂₃NO₂
MW: 285.81 g/mol
Melting Pt: 176 °C
MDL Number: MFCD06658143
CAS Number: 130198-05-9
https://us.vwr.com/stibo/web/std.lang.all/07/05/28930705.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR