2-Amino-3-thiophenecarbonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Aminothiophene-3-carbonitrile

A2488-5G
TCA2488-5GEA 61.87 USD
TCA2488-5G
2-Amino-3-thiophenecarbonitrile ≥98.0% (by GC)
2-Amino-3-thiophenecarbonitrile

CAS Number: 4651-82-5
MDL Number: MFCD00706298
Molecular Formula: C5H4N2S
Molecular Weight: 124.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 106


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2-Amino-3-thiophenecarbonitrile

Formula: C₅H₄N₂S
MW: 124.16 g/mol
Melting Pt: 106 °C
MDL Number: MFCD00706298
CAS Number: 4651-82-5
https://us.vwr.com/stibo/web/std.lang.all/06/85/28930685.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR