Aztreonam ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-[[(Z)-[1-(2-Amino-4-thiazolyl)-2-[[(2S,3S)-2-methyl-4-oxo-1-sulfo-3-azetidinyl]amino]-2-oxoethylidene]amino]oxy]-2-methylpropionic Acid

A2466-500MG A2466-5G
TCA2466-500MGEA 96.41 USD
TCA2466-500MG TCA2466-5G
Aztreonam ≥98.0% (by HPLC)
Aztreonam

CAS Number: 78110-38-0
MDL Number: MFCD00072145
Molecular Formula: C13H17N5O8S2
Molecular Weight: 435.43
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 227
Specific rotation [a]20/D: -28 deg (C=0.5, H2O)


Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: 2-[[(Z)-[1-(2-Amino-4-thiazolyl)-2-[[(2S,3S)-2-methyl-4-oxo-1-sulfo-3-azetidinyl]amino]-2-oxoethylidene]amino]oxy]-2-methylpropionic Acid

Formula: C₁₃H₁₇N₅O₈S₂
MW: 435.43 g/mol
Melting Pt: 227 °C
MDL Number: MFCD00072145
CAS Number: 78110-38-0
https://us.vwr.com/stibo/web/std.lang.all/06/56/28930656.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR