Azelnidipine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl Ester
CAS Number: 123524-52-7
MDL Number: MFCD00865803
Molecular Formula: C33H34N4O6
Molecular Weight: 582.66
Purity/Analysis Method:
Form: Crystal
Product Categories: Bioscience, Antihypertensive Ingredients, Voltage-dependent Calcium Channel (VDCC) Antagonists
Formula:
C₃₃H₃₄N₄O₆ MW: 582.66 g/mol |
MDL Number:
MFCD00865803 CAS Number: 123524-52-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...