Thiophene-2-methylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Thiophenemethylamine
, 2-(Aminomethyl)thiophene
A2419-25G
A2419-5G
TCA2419-25GEA
235
USD
TCA2419-25G
TCA2419-5G
Thiophene-2-methylamine ≥98.0% (by GC, titration analysis)
Thiophene-2-methylamine
CAS Number: 27757-85-3
MDL Number: MFCD00005460
Molecular Formula: C5H7NS
Molecular Weight: 113.18
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 74
Specific Gravity (20/20): 1.14
Other Product Names / Synonyms: (2-Thienyl)methylamine
Formula:
C₅H₇NS MW: 113.19 g/mol Boiling Pt: 99 °C Density: 1.14 (20 °C) Flash Pt: 74 °C |
MDL Number:
MFCD00005460 CAS Number: 27757-85-3 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...