N-(5-Aminopentyl)acetamide ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-Acetyl-1,5-pentanediamine
A2393-5G
TCA2393-5GEA
323.36
USD
TCA2393-5G
N-(5-Aminopentyl)acetamide ≥97.0% (by GC, titration analysis)
N-(5-Aminopentyl)acetamide
CAS Number: 32343-73-0
Molecular Formula: C7H16N2O
Molecular Weight: 144.22
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 110
Specific Gravity (20/20): 0.98
Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-Acetyl-1,5-pentanediamine
Formula:
C₇H₁₆N₂O MW: 144.22 g/mol Boiling Pt: 110 °C Density: 0.98 (20 °C) |
CAS Number:
32343-73-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...