Anastrozole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
alpha,alpha,alpha',alpha'-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile
CAS Number: 120511-73-1
MDL Number: MFCD00866298
Molecular Formula: C17H19N5
Molecular Weight: 293.37
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 84
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Inhibitors (for Pharmacological Research), Antitumor Ingredients
Other Product Names / Synonyms: alpha,alpha,alpha',alpha'-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile
Formula:
C₁₇H₁₉N₅ MW: 293.37 g/mol Melting Pt: 84 °C |
MDL Number:
MFCD00866298 CAS Number: 120511-73-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...