Acamprosate Calcium ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Calcium 3-Acetamido-1-propanesulfonate3-Acetamido-1-propanesulfonic Acid Calcium Salt

A2359-1G
TCA2359-1GEA 60.55 USD
TCA2359-1G
Acamprosate Calcium ≥98.0% (by HPLC, total nitrogen)
Acamprosate calcium

CAS Number: 77337-73-6
MDL Number: MFCD00886588
Molecular Formula: C5H11NO4S
Molecular Weight: 400.47
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 270


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: Calcium 3-Acetamido-1-propanesulfonate, 3-Acetamido-1-propanesulfonic Acid Calcium Salt

Formula: C₅H₁₁NO₄S
MW: 400.47 g/mol
Melting Pt: 270 °C
MDL Number: MFCD00886588
CAS Number: 77337-73-6
https://us.vwr.com/stibo/web/std.lang.all/05/50/28930550.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR