TCIAA2346

A2346-1G
TCA2346-1GEA 56.78 USD
TCA2346-1G
TCIAA2346
CAS Number: 48149-72-0
MDL Number: MFCD01320363
Molecular Formula: C9H16O6
Molecular Weight: 220.22
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Melting point (°C): 145
Specific rotation [a]20/D: 176 deg (C=1, H2O)
Storage Temperature: 0°C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR