3-Amino-3-(4-methylphenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
DL-3-Amino-3-p-tolyl-propionic acid
, 3-Amino-3-p-tolyl-propionic acid
A2345-25G
A2345-5G
TCA2345-25GEA
422
USD
TCA2345-25G
TCA2345-5G
3-Amino-3-(4-methylphenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-3-(4-methylphenyl)propionic acid
CAS Number: 68208-18-4
MDL Number: MFCD01863249
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 222
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...