2-Amino-6-fluorobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Cyano-3-fluoroaniline
A2344-5G
A2344-25G
TCA2344-25GEA
241
USD
TCA2344-25G
TCA2344-5G
2-Amino-6-fluorobenzonitrile ≥98.0% (by GC)
2-Amino-6-fluorobenzonitrile
CAS Number: 77326-36-4
MDL Number: MFCD00015477
Molecular Formula: C7H5FN2
Molecular Weight: 136.13
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 129
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Cyano-3-fluoroaniline
Formula:
C₇H₅FN₂ MW: 136.13 g/mol Melting Pt: 129 °C |
MDL Number:
MFCD00015477 CAS Number: 77326-36-4 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...