4-Pentylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4-n-Pentylbenzeneboronic acid(4-Pentylphenyl)boronic acid

A2336-5G
TCA2336-5GEA 59.85 USD
TCA2336-5G
4-Pentylphenylboronic acid (contains varying amounts of Anhydride)
4-Pentylphenylboronic acid

CAS Number: 121219-12-3
MDL Number: MFCD00995151
Molecular Formula: C11H17BO2
Molecular Weight: 192.07
Form: Crystal
Color: White
Melting point (°C): 84


Other Product Names / Synonyms: 4-Amylbenzeneboronic Acid, 4-Pentylphenylboronic Acid, 4-Pentylbenzeneboronic Acid

Formula: C₁₁H₁₇BO₂
MW: 192.07 g/mol
Melting Pt: 84 °C
MDL Number: MFCD00995151
CAS Number: 121219-12-3
https://us.vwr.com/stibo/web/std.lang.all/05/22/28930522.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR