(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (S)-(-)-1-(2-Amino-1-naphthyl)-2-naphthol(S)-(-)-NOBIN(S)-(-)-2'-Amino-1,1'-binaphthalen-2-ol

A2317-100MG
TCA2317-100MGEA 267 USD
TCA2317-100MG
(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl ≥98.0% (by HPLC)
(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl

CAS Number: 137848-29-4
MDL Number: MFCD01882346
Molecular Formula: C20H15NO
Molecular Weight: 285.35
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 173
Specific rotation [a]20/D: -121 deg (C=1, THF)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis
Other Product Names / Synonyms: (S)-(-)-1-(2-Amino-1-naphthyl)-2-naphthol, (S)-(-)-NOBIN, (S)-(-)-2'-Amino-1,1'-binaphthalen-2-ol

Formula: C₂₀H₁₅NO
MW: 285.35 g/mol
Melting Pt: 173 °C
MDL Number: MFCD01882346
CAS Number: 137848-29-4
https://us.vwr.com/stibo/web/std.lang.all/04/99/28930499.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR