N-(Allyloxycarbonyloxy)succinimide ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
Allyl N-Succinimidyl Carbonate
A2302-5G
A2302-25G
TCA2302-5GEA
98.32
USD
TCA2302-5G
TCA2302-25G
N-(Allyloxycarbonyloxy)succinimide ≥97.0% (by GC)
N-(Allyloxycarbonyloxy)succinimide
CAS Number: 135544-68-2
Molecular Formula: C8H9NO5
Molecular Weight: 199.16
Purity/Analysis Method:
Form: Crystal
Freezing point (°C): 21
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Monomers
Other Product Names / Synonyms: Allyl N-Succinimidyl Carbonate
Formula:
C₈H₉NO₅ MW: 199.16 g/mol |
MDL Number:
MFCD02684388 CAS Number: 135544-68-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...