Allyl(tert-butyl)dimethylsilane ≥97.0% (by GC)

Supplier: TCI America

A2298-5G
TCA2298-5GEA 188.72 USD
TCA2298-5G
Allyl(tert-butyl)dimethylsilane ≥97.0% (by GC)
Allyl(tert-butyl)dimethylsilane

CAS Number: 74472-22-3
Molecular Formula: C9H20Si
Molecular Weight: 156.34
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 152
Specific Gravity (20/20): 0.77

Formula: C₉H₂₀Si
MW: 156.34 g/mol
Boiling Pt: 152 °C
Density: 0.77 (20 °C)
MDL Number: MFCD18207712
CAS Number: 74472-22-3
https://us.vwr.com/stibo/web/std.lang.all/04/70/28930470.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR