11-Azido-3,6,9-trioxaundecanoic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Azido-PEG3-carboxylate
A2293-1G
TCA2293-1GEA
498.43
USD
TCA2293-1G
11-Azido-3,6,9-trioxaundecanoic acid ≥97.0% (by titrimetric analysis)
11-Azido-3,6,9-trioxaundecanoic acid
CAS Number: 172531-37-2
Molecular Formula: C8H15N3O5
Molecular Weight: 233.22
Purity/Analysis Method:
Form: Clear Liquid
Product Categories: Synthetic Organic Chemistry, Azide Compounds (Click Chemistry), Bioscience, Crosslinkers, PEGylation Reagents & PEG Spacers
Other Product Names / Synonyms: Azido-PEG3-carboxylate
Formula:
C₈H₁₅N₃O₅ MW: 233.22 g/mol |
CAS Number:
172531-37-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...