(1R,3S)-3-Aminocyclohexanecarboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A2287-200MG
TCA2287-200MGEA 453 USD
TCA2287-200MG
(1R,3S)-3-Aminocyclohexanecarboxylic acid ≥98.0% (by titrimetric analysis)
(1R,3S)-3-Aminocyclohexanecarboxylic acid

CAS Number: 81131-39-7
MDL Number: MFCD09753647
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 279
Specific rotation [a]20/D: -5 deg (C=1, H2O)

Formula: C₇H₁₃NO₂
Melting Pt: 279 °C
MDL Number: MFCD09753647
CAS Number: 81131-39-7
https://us.vwr.com/stibo/web/std.lang.all/04/56/28930456.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR