4-Amino-3-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-3-picoline3-Methylpyridin-4-amine3-Methyl-4-pyridylamine

A2283-1G
TCA2283-1GEA 59.89 USD
TCA2283-1G
4-Amino-3-methylpyridine ≥98.0% (by GC, titration analysis)
4-Amino-3-methylpyridine

CAS Number: 1990-90-5
MDL Number: MFCD01704431
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 108


Other Product Names / Synonyms: 4-Amino-3-picoline

Formula: C₆H₈N₂
MW: 108.14 g/mol
Melting Pt: 108 °C
MDL Number: MFCD01704431
CAS Number: 1990-90-5
https://us.vwr.com/stibo/web/std.lang.all/04/52/28930452.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR