2-Amino-4,6-dichlorophenol ≥98.0% (by titrimetric analysis)

Synonyms: 3,5-Dichloro-6-hydroxyaniline

A2246-25G A2246-5G
TCA2246-25GEA 175.38 USD
TCA2246-25G TCA2246-5G
2-Amino-4,6-dichlorophenol ≥98.0% (by titrimetric analysis)
2-Amino-4,6-dichlorophenol

CAS Number: 527-62-8
MDL Number: MFCD00035766
Molecular Formula: C6H5Cl2NO
Molecular Weight: 178.01
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 94


Other Product Names / Synonyms: 3,5-Dichloro-2-hydroxyaniline

Formula: C₆H₅Cl₂NO
MW: 178.01 g/mol
Melting Pt: 94 °C
MDL Number: MFCD00035766
CAS Number: 527-62-8
https://us.vwr.com/stibo/web/std.lang.all/04/23/28930423.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR