1-Allylimidazole ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Allyl-1H-imidazole

A2207-25G
TCA2207-25GEA 44.66 USD
TCA2207-25G
1-Allylimidazole ≥97.0% (by GC, titration analysis)
1-Allylimidazole

CAS Number: 31410-01-2
MDL Number: MFCD00037894
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 101
Specific Gravity (20/20): 1.01

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 116 °C
Density: 1.01 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00037894
CAS Number: 31410-01-2
https://us.vwr.com/stibo/web/std.lang.all/03/93/28930393.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR