4-Amino-2,5-difluorobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
A2106-1G
A2106-5G
TCA2106-5GEA
161
USD
TCA2106-5G
TCA2106-1G
4-Amino-2,5-difluorobenzonitrile ≥98.0% (by GC)
4-Amino-2,5-difluorobenzonitrile
CAS Number: 112279-61-5
MDL Number: MFCD00190102
Molecular Formula: C7H4F2N2
Molecular Weight: 154.12
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 97
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Cyano-2,5-difluoroaniline
Formula:
C₇H₄F₂N₂ MW: 154.12 g/mol Melting Pt: 97 °C |
MDL Number:
MFCD00190102 CAS Number: 112279-61-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...