Allyl methyl disulfide ≥90.0% (by GC)
Supplier: TCI America
A1939-5G
TCA1939-5GEA
145
USD
TCA1939-5G
Allyl methyl disulfide ≥90.0% (by GC)
Allyl methyl disulphide
CAS Number: 2179-58-0
MDL Number: MFCD00040023
Molecular Formula: C4H8S2
Molecular Weight: 120.23
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 35
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...