TATU (O-(7-aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: TATU

A1861-1G A1861-5G
TCA1861-1GEA 63 USD
TCA1861-1G TCA1861-5G
TATU (O-(7-aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥98.0% (by HPLC, total nitrogen)
TATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)

CAS Number: 873798-09-5
Molecular Formula: C10H15BF4N6O
Molecular Weight: 322.07
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 184
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Peptide Synthesis
Other Product Names / Synonyms: TATU

Formula: C₁₀H₁₅BF₄N₆O
MW: 322.1 g/mol
Melting Pt: 184 °C
CAS Number: 873798-09-5
https://us.vwr.com/stibo/web/std.lang.all/99/65/28929965.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR