Atrazine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine

A1650-5G
TCA1650-5GEA 132.02 USD
TCA1650-5G
Atrazine ≥97.0% (by GC, titration analysis)
Atrazine

CAS Number: 1912-24-9
MDL Number: MFCD00041810
Molecular Formula: C8H14ClN5
Molecular Weight: 215.69
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 177


Product Categories: Bioscience, Herbicide Ingredients
Other Product Names / Synonyms: 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine

Formula: C₈H₁₄ClN₅
MW: 215.69 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00041810
CAS Number: 1912-24-9
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/98/20/28929820.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR