Allylmagnesium chloride ∼11% ∼1.0 mol/L in THF
A1554-100G
TCA1554-100GEA
63.27
USD
TCA1554-100G
Allylmagnesium chloride ∼11% ∼1.0 mol/L in THF
Allylmagnesium chloride
CAS Number: 2622-05-1
MDL Number: MFCD00000473
Molecular Formula: C3H5ClMg
Molecular Weight: 100.83
Formula:
C₃H₅ClMg |
MDL Number:
MFCD00000473 CAS Number: 2622-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...