2-Bromo-5-(trifluoromethyl)aniline ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
3-Amino-4-bromo-α,α,α-trifluorotoluene
A1483-5G
TCA1483-5GEA
70
USD
TCA1483-5G
2-Bromo-5-(trifluoromethyl)aniline ≥97.0% (by GC)
2-Bromo-5-(trifluoromethyl)aniline
CAS Number: 454-79-5
MDL Number: MFCD00007637
Molecular Formula: C7H5BrF3N
Molecular Weight: 240.02
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 113
Specific Gravity (20/20): 1.71
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Bromo-5-trifluoromethylaniline
Formula:
C₇H₅BrF₃N MW: 240.02 g/mol Boiling Pt: 96 °C Density: 1.71 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00007637 CAS Number: 454-79-5 UN: 2810 ADR: 6.1,III |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...