(R)-(-)-2-Amino-1-butanol ≥94.0% (by GC)

Supplier: TCI America

Synonyms: L(-)-2-Amino-1-butanol

A0972-5ML
TCA0972-5MLEA 51.27 USD
TCA0972-5ML
(R)-(-)-2-Amino-1-butanol ≥94.0% (by GC)
(R)-(-)-2-Amino-1-butanol

CAS Number: 5856-63-3
MDL Number: MFCD00064419
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method: 94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 174
Flash Point (°C): 95
Specific Gravity (20/20): 0.95
Specific rotation [a]20/D: -9.5 deg (neat)

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 174 °C
Density: 0.95 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00064419
CAS Number: 5856-63-3
https://us.vwr.com/stibo/web/std.lang.all/87/22/28928722.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR