(S)-(+)-2-(Anilinomethyl)pyrrolidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (S)-N-(Pyrrolidin-2-ylmethyl)aniline

A0945-5G A0945-1G
TCA0945-5GEA 490.03 USD
TCA0945-5G TCA0945-1G
(S)-(+)-2-(Anilinomethyl)pyrrolidine ≥98.0% (by titrimetric analysis)
(S)-(+)-2-(Anilinomethyl)pyrrolidine

CAS Number: 64030-44-0
MDL Number: MFCD00015890
Molecular Formula: C11H16N2
Molecular Weight: 176.26
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 113
Freezing point (°C): 19
Specific Gravity (20/20): 1.05
Specific rotation [a]20/D: 18 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (S)-(+)-N-(2-Pyrrolidinomethyl)aniline

Formula: C₁₁H₁₆N₂
MW: 176.2581 g/mol
Boiling Pt: 126 °C
Density: 1.05 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00015890
CAS Number: 64030-44-0
https://us.vwr.com/stibo/web/std.lang.all/86/93/28928693.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR