Aminomethanesulfonic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A0852-25G
TCA0852-25GEA 29.62 USD
TCA0852-25G
Aminomethanesulfonic acid ≥97.0% (by titrimetric analysis)
Aminomethanesulphonic acid

CAS Number: 13881-91-9
MDL Number: MFCD00008124
Molecular Formula: CH5NO3S
Molecular Weight: 111.12
Purity/Analysis Method: 97.0% (T)
Form: Crystal

Formula: CH₅NO₃S
MW: 111.12 g/mol
MDL Number: MFCD00008124
CAS Number: 13881-91-9
https://us.vwr.com/stibo/web/std.lang.all/85/99/28928599.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR