Allyl mercaptan ≥80.0% (by GC)
Supplier: TCI America
A0777-25ML
TCA0777-25MLEA
136.08
USD
TCA0777-25ML
Allyl mercaptan ≥80.0% (by GC)
Allyl mercaptan
CAS Number: 870-23-5
MDL Number: MFCD00004894
Molecular Formula: C3H6S
Molecular Weight: 74.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 69
Flash Point (°C): 21
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Propene-1-thiol
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...