TCIAA0637

A0637-25G
TCA0637-25GEA 493.15 USD
TCA0637-25G
TCIAA0637
CAS Number: 1186-52-3
MDL Number: MFCD00051051
Molecular Formula: C2D4O2
Molecular Weight: 64.08
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 116
Melting point (°C): 16
Flash Point (°C): 40
Specific Gravity (20/20): 1.12

Other Product Names / Synonyms: Tetradeuteroacetic Acid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR