(±)-3-Amino-1,2-propanediol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (±)-3-Amino-1,2-dihydroxy propane3-Amino-1,2-propanediol

A0628-25G
TCA0628-25GEA 64.83 USD
TCA0628-25G
(±)-3-Amino-1,2-propanediol ≥98.0% (by GC, titration analysis)
(±)-3-Amino-1,2-propanediol

CAS Number: 616-30-8
MDL Number: MFCD00008140
Molecular Formula: C3H9NO2
Molecular Weight: 91.11
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 155

Formula: C₃H₉NO₂
MW: 91.11 g/mol
Flash Pt: 155 °C
MDL Number: MFCD00008140
CAS Number: 616-30-8
https://us.vwr.com/stibo/web/std.lang.all/80/43/28928043.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR