5-Amino-1-naphthol ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5,1-ANL

A0358-1G
TCA0358-1GEA 72.57 USD
TCA0358-1G
5-Amino-1-naphthol ≥97.0% (by HPLC, titration analysis)
5-Amino-1-naphthol

CAS Number: 83-55-6
MDL Number: MFCD00041826
Molecular Formula: C10H9NO
Molecular Weight: 159.19
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 193

Formula: C₁₀H₉NO
Melting Pt: 193 °C
MDL Number: MFCD00041826
CAS Number: 83-55-6
https://us.vwr.com/stibo/web/std.lang.all/77/97/28927797.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR