5-Amino-1-naphthalenesulfonic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
5-Aminonaphthalene-1-sulphonic acid
, 5-Amino-α-naphthalenesulphonic acid
, 5-Aminonaphthalene-1-sulfonic acid
, 5-Amino-α-naphthalenesulfonic acid
A0345-25G
TCA0345-25GEA
143.12
USD
TCA0345-25G
5-Amino-1-naphthalenesulfonic acid ≥98.0% (by titrimetric analysis)
5-Amino-1-naphthalenesulphonic acid
CAS Number: 84-89-9
MDL Number: MFCD00014315
Molecular Formula: C10H9NO3S
Molecular Weight: 223.25
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: Laurent's Acid, 1-Naphthylamine-5-sulfonic Acid
Formula:
C₁₀H₉NO₃S MW: 223.25 g/mol |
MDL Number:
MFCD00014315 CAS Number: 84-89-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...