TCIAA0340

A0340-5G
TCA0340-5GEA 136.93 USD
TCA0340-5G
TCIAA0340
CAS Number: 50976-35-7
MDL Number: MFCD00070485
Molecular Formula: C10H9NO6S2
Molecular Weight: 347.27
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C

Other Product Names / Synonyms: Amino J Acid Disodium Salt, 6-Amino-1,3-naphthalenedisulfonic Acid Disodium Salt, 2-Naphthylamine-5,7-disulfonic Acid Disodium Salt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR