Acridine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A0294-5G
TCA0294-5GEA 155.3 USD
TCA0294-5G
Acridine ≥98.0% (by HPLC, titration analysis)
Acridine

CAS Number: 260-94-6
MDL Number: MFCD00005025
Molecular Formula: C13H9N
Molecular Weight: 179.22
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 346
Melting point (°C): 108

Formula: C₁₃H₉N
Boiling Pt: 346 °C
Melting Pt: 108 °C
MDL Number: MFCD00005025
CAS Number: 260-94-6
UN: 2713
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/77/20/28927720.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR