N-Acetylthiourea ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
1-Acetylthiourea
A0117-25G
TCA0117-25GEA
34
USD
TCA0117-25G
N-Acetylthiourea ≥98.0% (by total nitrogen basis)
N-Acetylthiourea
CAS Number: 591-08-2
MDL Number: MFCD00004937
Molecular Formula: C3H6N2OS
Molecular Weight: 118.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 166
Formula:
C₃H₆N₂OS Melting Pt: 166 °C |
MDL Number:
MFCD00004937 CAS Number: 591-08-2 UN: 2811 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...