1-Acetyl-2-phenylhydrazine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-PhenylacetohydrazideN-Acetyl-N'-phenylhydrazine

A0107-25G
TCA0107-25GEA 32.48 USD
TCA0107-25G
1-Acetyl-2-phenylhydrazine ≥98.0% (by GC)
1-Acetyl-2-phenylhydrazine

CAS Number: 114-83-0
MDL Number: MFCD00008672
Molecular Formula: C8H10N2O
Molecular Weight: 150.18
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 130

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Melting Pt: 130 °C
MDL Number: MFCD00008672
CAS Number: 114-83-0
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/75/55/28927555.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR