Diethyl acetylenedicarboxylate ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
Diethyl but-2-yne-1,4-dioate
A0089-250ML
A0089-25ML
TCA0089-250MLEA
745.01
USD
TCA0089-250ML
TCA0089-25ML
Diethyl acetylenedicarboxylate ≥96.0% (by GC)
Diethyl acetylenedicarboxylate
CAS Number: 762-21-0
MDL Number: MFCD00009186
Molecular Formula: C8H10O4
Molecular Weight: 170.16
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 115
Melting point (°C): 1
Flash Point (°C): 94
Specific Gravity (20/20): 1.07
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Acetylenedicarboxylic Acid Diethyl Ester
Formula:
C₈H₁₀O₄ MW: 170.16 g/mol Boiling Pt: 115 °C Melting Pt: 1 °C Density: 1.07 (20 °C) Flash Pt: 94 °C |
MDL Number:
MFCD00009186 CAS Number: 762-21-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...