Acetylenedicarboxylic acid ≥90.0% (by HPLC)
Supplier: TCI America
A0088-25G
A0088-5G
TCA0088-25GEA
154.28
USD
TCA0088-25G
TCA0088-5G
Acetylenedicarboxylic acid ≥90.0% (by HPLC)
Acetylenedicarboxylic acid
CAS Number: 142-45-0
MDL Number: MFCD00004362
Molecular Formula: C4H2O4
Molecular Weight: 114.06
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 178
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Monomers
Other Product Names / Synonyms: 2-Butynedioic Acid
Formula:
C₄H₂O₄ Melting Pt: 178 °C |
MDL Number:
MFCD00004362 CAS Number: 142-45-0 UN: 2928 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...