Anti-IgG Polyclonal Antibody (DyLight® 650)

Supplier: Columbia Biosciences
D10-110-1MG
10159-202EA 515.63 USD
10159-202
Anti-IgG Polyclonal Antibody (DyLight® 650)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host:
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR