Anti-6xHis Mouse Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))

Supplier: Columbia Biosciences
D11-1711
10065-514EA 544.54 USD
10065-514
Anti-6xHis Mouse Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Antibodies
Type:
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR