Anti-GDF15 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
600-406-B07
RL600-406-B07EA 492.08 USD
RL600-406-B07
Anti-GDF15 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR