Anti-SOD2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-G13
RL600-401-G13EA 501.23 USD
RL600-401-G13
Anti-SOD2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Bovine, Dog, Chicken, Dosophila, Guinea Pig, Hamster, Monkey, Rabbit, Sheep, Xenopus, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR