Anti-CACNA1D Mouse Monoclonal Antibody [clone: S38-8]

Supplier: Rockland Immunochemicals
200-301-F10
RL200-301-F10EA 432.48 USD
RL200-301-F10
Anti-CACNA1D Mouse Monoclonal Antibody [clone: S38-8]
Antibodies
Type: Primary
Antigen: CACNA1D
Clonality: Monoclonal
Clone: S38-8
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR