Anti-LOX Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-H91 100-401-H91
RL100401H91EA 538.82 USD
RL100401H91 CARL100401H91
Anti-LOX Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR