Anti-pagA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-H55 100-401-H55
RL100401H55EA 538.82 USD
CARL100401H55 RL100401H55
Anti-pagA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: pagA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Bacillus anthracis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR