Anti-PPAPDC2 Rabbit Polyclonal Antibody

Supplier: ProSci
5517
89417-254EA 531.09 USD
89417-254
Anti-PPAPDC2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPAPDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR