Anti-Ly49G Mouse Monoclonal Antibody [clone: AT8]

Supplier: BioLegend
137402 137402
137402-BLEA 273.08 USD
137402-BL CA137402-BL
Anti-Ly49G Mouse Monoclonal Antibody [clone: AT8]
Antibodies
Purified anti-mouse Ly49G [AT8]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly49G
Clonality: Monoclonal
Clone: AT8
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR