Anti-ULK3 Mouse Monoclonal Antibody [clone: 1404CT148.89.25]

Supplier: Abcepta
AM8437C
10218-382EA 577.43 USD
10218-382
Anti-ULK3 Mouse Monoclonal Antibody [clone: 1404CT148.89.25]
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: ULK3
Clonality: Monoclonal
Clone: 1404CT148.89.25
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR